Anti-H2AFZ Rabbit Polyclonal Antibody

Lieferant: US Biological
140574 140574-100
USBI140574EA 469 EUR
USBI140574 USBI140574-100
Anti-H2AFZ Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500

Type: Primary
Antigen: H2AFZ
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR