Anti-N-Methylpurine DNA Glycosylase Rabbit Polyclonal Antibody

Lieferant: US Biological
141596
USBI141596EA 499 EUR
USBI141596
Anti-N-Methylpurine DNA Glycosylase Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500

Type: Primary
Antigen: N-Methylpurine DNA Glycosylase
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR