Anti-UDP Glucose Ceramide Glucosyltransferase Rabbit Polyclonal Antibody

Lieferant: US Biological
142720-50
USBI142720-50EA 456 EUR
USBI142720-50
Anti-UDP Glucose Ceramide Glucosyltransferase Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500

Type: Primary
Antigen: UDP Glucose Ceramide Glucosyltransferase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR