Anti-Aflatoxin Mouse Monoclonal Antibody [clone: 13k5]

Lieferant: US Biological
143039
USBI143039EA 935 EUR
USBI143039
Anti-Aflatoxin Mouse Monoclonal Antibody [clone: 13k5]
Antikörper
Type: Primary
Antigen: Aflatoxin (aflatoxin)
Clonality: monoclonal
Clone: 13k5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR