Anti-Aflatoxin B1/M1 Rat Monoclonal Antibody [clone: 13k7]

Lieferant: US Biological
143041
USBI143041EA 1190 EUR
USBI143041
Anti-Aflatoxin B1/M1 Rat Monoclonal Antibody [clone: 13k7]
Antikörper
Type: Primary
Antigen: Aflatoxin B1/M1 (aflatoxin B1/M1)
Clonality: monoclonal
Clone: 13k7
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR