Anti-Calprotectin Mouse Monoclonal Antibody

Lieferant: US Biological
143119 143119-100
USBI143119EA 391 EUR
USBI143119 USBI143119-100
Anti-Calprotectin Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Calprotectin
Clonality: Monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR