Anti-BD-4 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
143552
USBI143552EA 324 EUR
USBI143552
Anti-BD-4 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 0.25-1.0 ?g/ml

Type: Primary
Antigen: BD-4
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR