Anti-CCL11 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
143570-25
USBI143570-25EA 324 EUR
USBI143570-25
Anti-CCL11 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 0.25-1 ?g/ml

Type: Primary
Antigen: CCL11 (chemokine (C-C motif) ligand 11)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR