Anti-DDT Rabbit Polyclonal Antibody

Lieferant: US Biological
144441
USBI144441EA 587 EUR
USBI144441
Anti-DDT Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DDT (D-dopachrome tautomerase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR