Anti-FABP5 Rabbit Polyclonal Antibody

Lieferant: US Biological
144502
USBI144502EA 587 EUR
USBI144502
Anti-FABP5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FABP5 (fatty acid binding protein 5 (psoriasis-associated))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR