Anti-FASLG Rabbit Polyclonal Antibody

Lieferant: US Biological
144511
USBI144511EA 467 EUR
USBI144511
Anti-FASLG Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FASLG (Fas ligand (TNF superfamily, member 6))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR