Anti-LAT Rabbit Polyclonal Antibody

Lieferant: US Biological
144835
USBI144835EA 467 EUR
USBI144835
Anti-LAT Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LAT (linker for activation of T cells)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR