Anti-NEB Mouse Monoclonal Antibody [clone: Neb-20]

Lieferant: US Biological
144950
USBI144950EA 587 EUR
USBI144950
Anti-NEB Mouse Monoclonal Antibody [clone: Neb-20]
Antikörper
Type: Primary
Antigen: NEB (nebulin)
Clonality: monoclonal
Clone: Neb-20
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR