Anti-p19INK4d Mouse Monoclonal Antibody [clone: IMD-19]

Lieferant: US Biological
145020
USBI145020EA 587 EUR
USBI145020
Anti-p19INK4d Mouse Monoclonal Antibody [clone: IMD-19]
Antikörper
Type: Primary
Antigen: p19INK4d
Clonality: monoclonal
Clone: IMD-19
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR