Anti-Pulmonary surfactant-associated protein A1 Rabbit Polyclonal Antibody

Lieferant: US Biological
145112
USBI145112EA 587 EUR
USBI145112
Anti-Pulmonary surfactant-associated protein A1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Pulmonary surfactant-associated protein A1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR