Anti-ROR1 Rabbit Polyclonal Antibody

Lieferant: US Biological
145134
USBI145134EA 391 EUR
USBI145134
Anti-ROR1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ROR1 (receptor tyrosine kinase-like orphan receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR