Anti-RPS6KA1 Rabbit Polyclonal Antibody

Lieferant: US Biological
145136
USBI145136EA 587 EUR
USBI145136
Anti-RPS6KA1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RPS6KA1 (ribosomal protein S6 kinase, 90kDa, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR