Anti-RRM2B Rabbit Polyclonal Antibody

Lieferant: US Biological
145137
USBI145137EA 587 EUR
USBI145137
Anti-RRM2B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RRM2B (ribonucleotide reductase M2 B (TP53 inducible))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR