Anti-Smad 1,2,3,5 Rabbit Polyclonal Antibody

Lieferant: US Biological
145193
USBI145193EA 587 EUR
USBI145193
Anti-Smad 1,2,3,5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Smad 1,2,3,5
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR