Anti-TEC Rabbit Polyclonal Antibody

Lieferant: US Biological
145254
USBI145254EA 587 EUR
USBI145254
Anti-TEC Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TEC (tec protein tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR