Anti-EOMES Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 644730]

Lieferant: US Biological
145453
USBI145453EA 790 EUR
USBI145453
Anti-EOMES Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 644730]
Antikörper
Do not freeze!

Type: Primary
Antigen: EOMES (eomesodermin)
Clonality: monoclonal
Clone: 644730
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR