Anti-FABP4 Rat Monoclonal Antibody [clone: 804914]

Lieferant: US Biological
145461
USBI145461EA 817 EUR
USBI145461
Anti-FABP4 Rat Monoclonal Antibody [clone: 804914]
Antikörper
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: FABP4 (fatty acid binding protein 4, adipocyte)
Clonality: monoclonal
Clone: 804914
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR