Anti-MARCO Sheep Polyclonal Antibody

Lieferant: US Biological
145517
USBI145517EA 924 EUR
USBI145517
Anti-MARCO Sheep Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.25 ?g/ml

Type: Primary
Antigen: MARCO (macrophage receptor with collagenous structure)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR