Anti-Semaphorin 4F Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 780225]

Lieferant: US Biological
145603
USBI145603EA 694 EUR
USBI145603
Anti-Semaphorin 4F Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 780225]
Antikörper
Do not freeze!

Type: Primary
Antigen: Semaphorin 4F
Clonality: monoclonal
Clone: 780225
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR