Anti-West Nile Virus, Matrix Rabbit Polyclonal Antibody

Lieferant: US Biological
146024
USBI146024EA 647 EUR
USBI146024
Anti-West Nile Virus, Matrix Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: West Nile Virus, Matrix
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR