Anti-West Nile Virus, Envelope Rabbit Polyclonal Antibody
Lieferant: US Biological
146030
USBI146030EA 647 EUR
USBI146030
Anti-West Nile Virus, Envelope Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1 ?g/ml
Type: Primary
Antigen: West Nile Virus, Envelope
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen