Anti-TIM-4 Rabbit Polyclonal Antibody

Lieferant: US Biological
146091
USBI146091EA 647 EUR
USBI146091
Anti-TIM-4 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 10 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: TIM-4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR