Anti-CLU Rabbit Polyclonal Antibody

Lieferant: US Biological
146099
USBI146099EA 647 EUR
USBI146099
Anti-CLU Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-1 ?g/ml; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: CLU (clusterin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR