Anti-PTK7 Rabbit Polyclonal Antibody

Lieferant: US Biological
146194
USBI146194EA 647 EUR
USBI146194
Anti-PTK7 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1 ?g/ml; Immunofluorescence: 20 ?g/ml; Immunochemistry: 2.5 ?g/ml

Type: Primary
Antigen: PTK7 (protein tyrosine kinase 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR