Anti-UNC93B1 Rabbit Polyclonal Antibody

Lieferant: US Biological
146259
USBI146259EA 574 EUR
USBI146259
Anti-UNC93B1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-1 ?g/ml; Immunofluorescence: 20 ?g/ml; Immunochemistry: 2.5 ?g/ml

Type: Primary
Antigen: UNC93B1 (unc-93 homolog B1 (C. elegans))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR