Anti-UBA2 Rabbit Polyclonal Antibody

Lieferant: US Biological
146657
USBI146657EA 574 EUR
USBI146657
Anti-UBA2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.25-0.5 ?g/ml

Type: Primary
Antigen: UBA2 (ubiquitin-like modifier activating enzyme 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR