Anti-NLRP4 Rabbit Polyclonal Antibody

Lieferant: US Biological
146720
USBI146720EA 647 EUR
USBI146720
Anti-NLRP4 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: NLRP4 (NLR family, pyrin domain containing 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR