Anti-LRRFIP1 Rabbit Polyclonal Antibody

Lieferant: US Biological
146814
USBI146814EA 647 EUR
USBI146814
Anti-LRRFIP1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: LRRFIP1 (leucine rich repeat (in FLII) interacting protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR