Anti-CHN2 Goat Polyclonal Antibody

Lieferant: US Biological
149880
USBI149880EA 809 EUR
USBI149880
Anti-CHN2 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:8000

Type: Primary
Antigen: CHN2 (chimerin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR