Anti-ATG12 Rabbit Polyclonal Antibody

Lieferant: US Biological
149987
USBI149987EA 647 EUR
USBI149987
Anti-ATG12 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-1 ?g/ml; Immunochemistry: 2.5 ?g/ml

Type: Primary
Antigen: ATG12 (autophagy related 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR