Anti-ANXA2 Goat Polyclonal Antibody

Lieferant: US Biological
149995
USBI149995EA 809 EUR
USBI149995
Anti-ANXA2 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 0.5-3?g/ml

Type: Primary
Antigen: ANXA2 (annexin A2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR