Anti-Creatine Kinase BB Isoenzyme Goat Polyclonal Antibody

Lieferant: US Biological
150196
USBI150196EA 688 EUR
USBI150196
Anti-Creatine Kinase BB Isoenzyme Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:2000; Western Blot: 0.1-0. 3?g/ml

Type: Primary
Antigen: Creatine Kinase BB Isoenzyme
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR