Anti-ERG Goat Polyclonal Antibody

Lieferant: US Biological
150379
USBI150379EA 809 EUR
USBI150379
Anti-ERG Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:16000; Immunochemistry: 4-8?g/ml

Type: Primary
Antigen: ERG (v-ets avian erythroblastosis virus E26 oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR