Anti-EXO1 Goat Polyclonal Antibody

Lieferant: US Biological
150384
USBI150384EA 809 EUR
USBI150384
Anti-EXO1 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:64000; Western Blot: 0.2-0.5 ?g/ml

Type: Primary
Antigen: EXO1 (exonuclease 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR