Anti-LCN2 Goat Polyclonal Antibody

Lieferant: US Biological
150616
USBI150616EA 809 EUR
USBI150616
Anti-LCN2 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:16000; Western Blot: 0. 01-0. 03?g/ml; Immunochemistry: 2-3?g/ml

Type: Primary
Antigen: LCN2 (lipocalin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR