Anti-MAML1 Goat Polyclonal Antibody

Lieferant: US Biological
150633
USBI150633EA 809 EUR
USBI150633
Anti-MAML1 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:128000; Western Blot: 0.1-0. 3?g/ml; Immunochemistry: 2.5-3. 8?g/ml

Type: Primary
Antigen: MAML1 (mastermind-like 1 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR