Anti-MTR Goat Polyclonal Antibody

Lieferant: US Biological
150673
USBI150673EA 809 EUR
USBI150673
Anti-MTR Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:32000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: MTR (5-methyltetrahydrofolate-homocysteine methyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR