Anti-POU5F1 Goat Polyclonal Antibody

Lieferant: US Biological
150738
USBI150738EA 809 EUR
USBI150738
Anti-POU5F1 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:16000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: POU5F1 (POU class 5 homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR