Anti-PPY Goat Polyclonal Antibody

Lieferant: US Biological
150838
USBI150838EA 748 EUR
USBI150838
Anti-PPY Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:32000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: PPY (pancreatic polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR