Anti-GRIN2D Goat Polyclonal Antibody

Lieferant: US Biological
151225
USBI151225EA 563 EUR
USBI151225
Anti-GRIN2D Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:32000

Type: Primary
Antigen: GRIN2D (glutamate receptor, ionotropic, N-methyl D-aspartate 2D)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR