Anti-Estriol Mouse Monoclonal Antibody

Lieferant: US Biological
151753
USBI151753EA 552 EUR
USBI151753
Anti-Estriol Mouse Monoclonal Antibody
Antikörper
  • Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500

Type: Primary
Antigen: Estriol
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR