Anti-Semaphorin 5B Mouse Monoclonal Antibody
Lieferant: US Biological
152103
USBI152103EA 556 EUR
USBI152103
Anti-Semaphorin 5B Mouse Monoclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500
Type: Primary
Antigen: Semaphorin 5B
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human