Anti-FABP4 Goat Polyclonal Antibody

Lieferant: US Biological
167401
USBI167401EA 601 EUR
USBI167401
Anti-FABP4 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:128000; Western Blot: 0. 03-0.1 ?g/ml

Type: Primary
Antigen: FABP4 (fatty acid binding protein 4, adipocyte)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR