Anti-GGT1 Goat Polyclonal Antibody

Lieferant: US Biological
167416
USBI167416EA 601 EUR
USBI167416
Anti-GGT1 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:128000; Western Blot: 0.1-0. 3?g/ml

Type: Primary
Antigen: GGT1 (gamma-glutamyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR