Anti-PLK5 Goat Polyclonal Antibody

Lieferant: US Biological
167478
USBI167478EA 640 EUR
USBI167478
Anti-PLK5 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:64000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: PLK5 (polo-like kinase 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR